Please login to your online account to display your discounted pricing

anti-Histone H4 (Monomethyl Lys20), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB212088

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB212088

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Gene Accession No. P62805
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Primary or Secondary Primary
Gene ID (Entrez) 121504
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Concentration 1.12mg/mL
Format Affinity Purified
Quantity 0.05mg
Host Species Rabbit
Immunogen Synthetic monomethylated peptide surrounding Lysine 20 of human Histone H4 [Swiss Prot P62805].
Cross Reactivity C. elegans
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Histone H4 (Monomethyl Lys20)
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal