Please login to your online account to display your discounted pricing

anti-Histone H4 (Monomethyl Arg3), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB212011

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB212011

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 121504
Formulation PBS, 30% glycerol
Immunogen Synthetic monomethylated peptide surrounding Arginine 3 of human Histone H4 [Swiss Prot P62805].
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human, mouse and rat. Predicted to react with most mammal species.
Primary or Secondary Primary
Gene Accession No. P62805
Concentration 0.43mg/mL
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Antigen Histone H4 (Monomethyl Arg3)
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Quantity 0.05mg