Please login to your online account to display your discounted pricing

anti-Histone H4 (Dimethyl Lys20), Polyclonal, Novus Biologicals™

$219.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic dimethylated peptide surrounding Lysine 20 of human Histone H4 [Swiss Prot P62805].
Antigen Histone H4 (Dimethyl Lys20)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB212089

 
novus biologicals canada
NB212089
Unlabeled 0.05mg Each for $384.00

NB212089SS

 
novus biologicals canada
NB212089SS
Unlabeled 0.025mg Each for $219.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic dimethylated peptide surrounding Lysine 20 of human Histone H4 [Swiss Prot P62805].
Antigen Histone H4 (Dimethyl Lys20)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Gene Accession No. P62805
Gene ID (Entrez) 121504
Formulation PBS, 30% glycerol
Concentration 0.53mg/mL