Please login to your online account to display your discounted pricing

anti-Histone H4 (ac Lys8), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB212044

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB212044

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P62805
Gene ID (Entrez) 121504
Conjugate Unlabeled
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Formulation PBS, 30% glycerol
Concentration 0.85mg/mL
Primary or Secondary Primary
Format Affinity Purified
Antigen Histone H4 (ac Lys8)
Cross Reactivity C. elegans
Cross Reactivity Human
Cross Reactivity Mouse
Regulatory Status RUO
Host Species Rabbit
Quantity 0.05mg
Monoclonal or Polyclonal Polyclonal
Immunogen Synthetic acetylated peptide surrounding lysine 8 of human histone H4 [Swiss Prot P62805].