Please login to your online account to display your discounted pricing

anti-Histone H4 (ac Lys12), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB212064SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB212064SS

  • / Each
 

Description & Specifications

Specifications

Formulation 0.02M Potassium phosphate, 0.15M Sodium Chloride, pH 7.2
Gene ID (Entrez) 121504
Concentration 0.69mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Accession No. P62805
Test Specificity Human, mouse and rat. Predicted to react with most mammal species.
Host Species Rabbit
Isotype IgG
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Antigen Histone H4 (ac Lys12)
Immunogen Histone H4
Format Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mg