Please login to your online account to display your discounted pricing

anti-Histone H4 (ac Lys12), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB212064

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB212064

  • / Each
 

Description & Specifications

Specifications

Antigen Histone H4 (ac Lys12)
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Concentration 0.69mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation 0.02M Potassium phosphate, 0.15M Sodium Chloride, pH 7.2
Gene Accession No. P62805
Host Species Rabbit
Immunogen Histone H4
Isotype IgG
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 121504
Test Specificity Human, mouse and rat. Predicted to react with most mammal species.
Target Species Human
Target Species Mammalia
Target Species Murine
Target Species Rat