Please login to your online account to display your discounted pricing

anti-Histone H4 (ac Lys12), Polyclonal, Novus Biologicals™

$219.00 - $384.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Histone H4
Antigen Histone H4 (ac Lys12)
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB212064

 
novus biologicals canada
NB212064
Unlabeled 0.05mg Each for $384.00

NB212064SS

 
novus biologicals canada
NB212064SS
Unlabeled 0.025mg Each for $219.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Histone H4
Antigen Histone H4 (ac Lys12)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Test Specificity Human, mouse and rat. Predicted to react with most mammal species.
Gene Accession No. P62805
Gene ID (Entrez) 121504
Formulation 0.02M Potassium phosphate, 0.15M Sodium Chloride, pH 7.2
Concentration 0.69mg/mL