Please login to your online account to display your discounted pricing

anti-Histone H3 (Trimethyl Lys9), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB211073SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB211073SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Gene Accession No. Q71DI3
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Concentration 0.85mg/mL
Cross Reactivity C. elegans
Cross Reactivity Human
Cross Reactivity Mouse
Formulation PBS, 30% glycerol
Gene ID (Entrez) 126961
Conjugate Unlabeled
Primary or Secondary Primary
Quantity 0.025mg
Antigen Histone H3 (Trimethyl Lys9)
Immunogen Synthetic trimethylated peptide surrounding Lysine 9 of human Histone H3.2 [Swiss Prot Q71DI3].
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Regulatory Status RUO
Host Species Rabbit