Please login to your online account to display your discounted pricing

anti-Histone H3 (Trimethyl Lys9), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB211073

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB211073

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 126961
Quantity 0.05mg
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Cross Reactivity C. elegans
Cross Reactivity Human
Cross Reactivity Mouse
Gene Accession No. Q71DI3
Concentration 0.85mg/mL
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Regulatory Status RUO
Format Affinity Purified
Host Species Rabbit
Antigen Histone H3 (Trimethyl Lys9)
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Immunogen Synthetic trimethylated peptide surrounding Lysine 9 of human Histone H3.2 [Swiss Prot Q71DI3].