Please login to your online account to display your discounted pricing

anti-Histone H3 (Trimethyl Lys36), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB211253SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB211253SS

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Gene Accession No. Q71DI3
Monoclonal or Polyclonal Polyclonal
Concentration 0.59mg/mL
Host Species Rabbit
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Cross Reactivity C. elegans
Cross Reactivity Human
Cross Reactivity Mouse
Primary or Secondary Primary
Conjugate Unlabeled
Immunogen Synthetic trimethylated peptide surrounding Lysine 36 of human Histone H3.2 [Swiss Prot Q71DI3].
Gene ID (Entrez) 126961
Format Affinity Purified
Applications Dot Blot
Applications Western Blotting
Quantity 0.025mg
Antigen Histone H3 (Trimethyl Lys36)
Regulatory Status RUO