Please login to your online account to display your discounted pricing

anti-Histone H3 (Monomethyl Lys79), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB211381SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB211381SS

  • / Each
 

Description & Specifications

Specifications

Antigen Histone H3 (Monomethyl Lys79)
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q71DI3
Host Species Rabbit
Immunogen Synthetic monomethylated peptide surrounding Lysine 79 of human Histone H3.2 [Swiss Prot Q71DI3].
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 126961
Test Specificity Human, mouse, and monkey. Predicted to react with many species including S. cerevisiae, rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Target Species Human
Target Species Murine
Target Species Primate