Please login to your online account to display your discounted pricing

anti-Histone H3 (Monomethyl Lys79), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB211381

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB211381

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 126961
Test Specificity Human, mouse, and monkey. Predicted to react with many species including S. cerevisiae, rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Gene Accession No. Q71DI3
Conjugate Unlabeled
Antigen Histone H3 (Monomethyl Lys79)
Formulation PBS, 30% glycerol
Primary or Secondary Primary
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Quantity 0.05mg
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Immunogen Synthetic monomethylated peptide surrounding Lysine 79 of human Histone H3.2 [Swiss Prot Q71DI3].
Format Affinity Purified
Regulatory Status RUO