missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Histone H3 (Monomethyl Lys18), Polyclonal, Novus Biologicals™

$219.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic monomethylated peptide surrounding Lysine 18 of human Histone H3.2 [Swiss Prot Q71DI3].
Antigen Histone H3 (Monomethyl Lys18)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB211141 Novus Biologicals Canada
NB211141
0.05mg Each for $384.00
NB211141SS Novus Biologicals Canada
NB211141SS
0.025mg Each for $219.00
Specifications Description & Specifications

Specifications

Rabbit
Histone H3 (Monomethyl Lys18)
Affinity Purified
RUO
Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
126961
0.64mg/mL
Synthetic monomethylated peptide surrounding Lysine 18 of human Histone H3.2 [Swiss Prot Q71DI3].
Polyclonal
Primary
Chromatin Immunoprecipitation,Dot Blot,Immunocytochemistry,Immunofluorescence,Western Blot
Q71DI3
PBS, 30% glycerol