Please login to your online account to display your discounted pricing

anti-Histone H3 (Dimethyl Lys9), Polyclonal, Novus Biologicals™

$219.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic dimethylated peptide surrounding Lysine 9 of human Histone H3.2 [Swiss Prot Q71DI3].
Antigen Histone H3 (Dimethyl Lys9)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB211072SS

 
novus biologicals canada
NB211072SS
Unlabeled 0.025mg Each for $219.00

NB211072

 
novus biologicals canada
NB211072
Unlabeled 0.05mg Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic dimethylated peptide surrounding Lysine 9 of human Histone H3.2 [Swiss Prot Q71DI3].
Antigen Histone H3 (Dimethyl Lys9)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Gene Accession No. Q71DI3
Gene ID (Entrez) 126961
Formulation PBS, 30% glycerol
Concentration 0.6mg/mL