Please login to your online account to display your discounted pricing

anti-Histone H3 (Dimethyl Lys37), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB211272SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB211272SS

  • / Each
 

Description & Specifications

Specifications

Antigen Histone H3 (Dimethyl Lys37)
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 0.39mg/mL
Conjugate Unlabeled
Cross Reactivity C. elegans
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q71DI3
Host Species Rabbit
Immunogen Synthetic dimethylated peptide surrounding Lysine 37 of human Histone H3 [Swiss Prot Q71DI3].
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 126961
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Target Species C. elegans
Target Species Human
Target Species Murine