Please login to your online account to display your discounted pricing

anti-Histone H3 (Dimethyl Lys37), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB211272

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB211272

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Conjugate Unlabeled
Gene Accession No. Q71DI3
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Format Affinity Purified
Gene ID (Entrez) 126961
Concentration 0.39mg/mL
Cross Reactivity C. elegans
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen Synthetic dimethylated peptide surrounding Lysine 37 of human Histone H3 [Swiss Prot Q71DI3].
Primary or Secondary Primary
Quantity 0.05mg
Regulatory Status RUO
Antigen Histone H3 (Dimethyl Lys37)