Please login to your online account to display your discounted pricing

anti-Histone H3, Clone: CMA301, EMD Millipore™


Manufacturer: emd millipore corporation  051341

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 05-134-1MI

  • / Each
 

Description & Specifications

Specifications

Immunogen Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, conjugated to KLH.
Isotype κ
Format Affinity Purified
Formulation Mouse monoclonal IgG2bκ supernatant in PBS with 0.05% sodium azide.
Gene Symbols H3.4, H3/g , H3/t, H3FT, H3T, H3t, MGC126886, MGC126888, OTTHUMP00000037945
Host Species Mouse
Applications Dot Blot
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Peptide affinity chromatography
Quantity 100μg
Storage Requirements 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Vertebrate
Antigen Histone H3
Gene Accession No. NM_003493
Monoclonal or Polyclonal Monoclonal
Clone CMA301
Conjugate Unlabeled