Please login to your online account to display your discounted pricing

anti-Histone H3 (ac Lys9), Polyclonal, Novus Biologicals™

$219.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic acetylated peptide surrounding Lysine 9 of human Histone H3.2 [Swiss Prot Q71DI3].
Antigen Histone H3 (ac Lys9)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB211074

 
novus biologicals canada
NB211074
Unlabeled 0.05mg Each for $384.00

NB211074SS

 
novus biologicals canada
NB211074SS
Unlabeled 0.025mg Each for $219.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic acetylated peptide surrounding Lysine 9 of human Histone H3.2 [Swiss Prot Q71DI3].
Antigen Histone H3 (ac Lys9)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant (Nicotiana tabacum, Zea mays, Triticum aestivum, and Encephalartos caffer) based on 100% sequence homology.
Gene Accession No. Q71DI3
Gene ID (Entrez) 126961
Formulation PBS, 30% glycerol
Concentration 0.68mg/mL