Please login to your online account to display your discounted pricing

anti-Histone H3 (ac Lys9, p Ser10), Polyclonal, Novus Biologicals™

$219.00 - $384.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Histone H3
Antigen Histone H3 (ac Lys9, p Ser10)
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB211075SS

 
novus biologicals canada
NB211075SS
Unlabeled 0.025mg Each for $219.00

NB211075

 
novus biologicals canada
NB211075
Unlabeled 0.05mg Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Histone H3
Antigen Histone H3 (ac Lys9, p Ser10)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Gene Accession No. Q71DI3
Gene ID (Entrez) 126961
Formulation 0.02M Potassium phosphate, 0.15M Sodium Chloride, pH 7.2
Concentration 0.25mg/mL