Please login to your online account to display your discounted pricing

anti-Histone H3 (ac Lys9, p Ser10), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB211075SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB211075SS

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Formulation 0.02M Potassium phosphate, 0.15M Sodium Chloride, pH 7.2
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Concentration 0.25mg/mL
Conjugate Unlabeled
Gene ID (Entrez) 126961
Gene Accession No. Q71DI3
Host Species Rabbit
Regulatory Status RUO
Cross Reactivity C. elegans
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.025mg
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen Histone H3
Format Affinity Purified
Antigen Histone H3 (ac Lys9, p Ser10)
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary