Please login to your online account to display your discounted pricing

anti-Histone H3 (ac Lys23), Polyclonal, Novus Biologicals™

$142.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Synthetic acetylated peptide surrounding Lysine 23 of human Histone H3 [Swiss Prot Q71DI3].
Antigen Histone H3 (ac Lys23)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB211164SS

 
novus biologicals canada
NB211164SS
Unlabeled 0.025mg Each for $142.00

NB211164

 
novus biologicals canada
NB211164
Unlabeled 0.05mg Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic acetylated peptide surrounding Lysine 23 of human Histone H3 [Swiss Prot Q71DI3].
Antigen Histone H3 (ac Lys23)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Dot Blot
Applications Western Blot
Test Specificity Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology.
Gene Accession No. Q71DI3
Gene ID (Entrez) 126961
Formulation PBS, 30% glycerol
Concentration 1.07mg/mL