Please login to your online account to display your discounted pricing

anti-Histone H3.3, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human Histone H3.3 was used as the immunogen for the antibody.
Antigen Histone H3.3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224697SS

 
novus biologicals canada
NBP224697SS
Unlabeled 0.025mg Each for $139.00

NBP224697

 
novus biologicals canada
NBP224697
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human Histone H3.3 was used as the immunogen for the antibody.
Antigen Histone H3.3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The Histone H3.3 amino acid sequence used as immunogen for the antibody is 100% homologous in human (Histone H3.3A, B, and C), Drosophila, orangutan, horse, oppossum, mouse, rat, cow, chicken, Xenopus, salmon and zebrafish.
Gene ID (Entrez) 3020
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL