Please login to your online account to display your discounted pricing

anti-Histone H2B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056633SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056633T

  • / Each
 

Description & Specifications

Specifications

Antigen Histone H2B
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Immunogen Amino acids 1-15 (MPDPAKSAPAPKKGS) of human Histone H2B were used as the immunogen.
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 440689
Test Specificity In PBMC, a 15 kDa band is observed.
Target Species Human