Please login to your online account to display your discounted pricing

anti-Histone H2B, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056633

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056633

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Test Specificity In PBMC, a 15 kDa band is observed.
Host Species Rabbit
Primary or Secondary Primary
Gene ID (Entrez) 440689
Formulation PBS containing 0.05% BSA
Format Purified
Quantity 0.2mL
Cross Reactivity Human
Antigen Histone H2B
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Immunogen Amino acids 1-15 (MPDPAKSAPAPKKGS) of human Histone H2B were used as the immunogen.
Applications Western Blotting