Please login to your online account to display your discounted pricing

anti-Histone H2B, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 1-15 (MPDPAKSAPAPKKGS) of human Histone H2B were used as the immunogen.
Antigen Histone H2B
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056633

 
novus biologicals canada
NB10056633
Unlabeled 0.2mL Each for $255.80

NB10056633T

 
novus biologicals canada
NB10056633SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 1-15 (MPDPAKSAPAPKKGS) of human Histone H2B were used as the immunogen.
Antigen Histone H2B
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In PBMC, a 15 kDa band is observed.
Gene ID (Entrez) 440689
Formulation PBS containing 0.05% BSA