Please login to your online account to display your discounted pricing

anti-Histone H2B, member S, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056347

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056347

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Primary or Secondary Primary
Conjugate Unlabeled
Test Specificity A 15 kDa band is observed. This antibody is routinely quality controlled in human lysates. Reacts with Chicken, Dog, Drosophila, Human, Most Mammals, Mouse, Rat and Zebrafish. his peptide sequence is 100% conserved between multiple species including human, mouse, rat, dog, chimpanzee, cow, chicken, duck, zebrafish, and drosophila.
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Concentration 0.5mg/mL
Gene ID (Entrez) 54145
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Drosophila
Cross Reactivity Human
Cross Reactivity Mammalia
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Zebrafish
Format Purified
Host Species Rabbit
Antigen Histone H2B, member S
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acid residues 111-125 AVSEGTKAVTKYTSS of human Histone H2B; GenBank no. CAI19747.1.
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mg