Please login to your online account to display your discounted pricing

anti-Histone Deacetylase 8/HDAC8 (p Ser39), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203991

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203991

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_060956
Formulation Rabbit IgG in phosphate buffered saline (without Mg2+, Ca2+), pH 7.4, 150mM NaCl and 50% glycerol.
Primary or Secondary Primary
Quantity 0.1mg
Gene ID (Entrez) 55869
Conjugate Unlabeled
Antigen Histone Deacetylase 8/HDAC8 (p Ser39)
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen This antibody was generated by immunizing rabbits with a synthetic non-phosphopeptide corresponding to human HDAC8 around the phosphorylation site of serine 39 (R-A-SP-M-V).
Cross Reactivity Human
Regulatory Status RUO
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified