Please login to your online account to display your discounted pricing

anti-Histone Deacetylase 8/HDAC8 (p Ser39), Polyclonal, Novus Biologicals™

$429.00 - $429.00

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic non-phosphopeptide corresponding to human HDAC8 around the phosphorylation site of serine 39 (R-A-SP-M-V).
Antigen Histone Deacetylase 8/HDAC8 (p Ser39)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203991

 
novus biologicals canada
NBP203991
Unlabeled 0.1mg Each for $429.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic non-phosphopeptide corresponding to human HDAC8 around the phosphorylation site of serine 39 (R-A-SP-M-V).
Antigen Histone Deacetylase 8/HDAC8 (p Ser39)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. NP_060956
Gene ID (Entrez) 55869
Formulation Rabbit IgG in phosphate buffered saline (without Mg2+, Ca2+), pH 7.4, 150mM NaCl and 50% glycerol.