Please login to your online account to display your discounted pricing

anti-Histone Deacetylase 2/HDAC2, Polyclonal, Novus Biologicals™

$142.00 - $409.37

Specifications

Host Species Rabbit
Immunogen Peptide sequence around aa. 392~396 (E-D-S-G-D) derived from Human HDAC2.
Antigen Histone Deacetylase 2/HDAC2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203980

 
novus biologicals canada
NBP203980
Unlabeled 0.1mg Each for $409.37

NBP203980SS

 
novus biologicals canada
NBP203980SS
Unlabeled 0.025mg Each for $142.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Peptide sequence around aa. 392~396 (E-D-S-G-D) derived from Human HDAC2.
Antigen Histone Deacetylase 2/HDAC2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Gene Accession No. NP_478057
Gene ID (Entrez) 3066
Formulation phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol.