Please login to your online account to display your discounted pricing

anti-Histone Deacetylase 2/HDAC2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203980SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203980SS

  • / Each
 

Description & Specifications

Specifications

Antigen Histone Deacetylase 2/HDAC2
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol.
Gene Accession No. NP_478057
Host Species Rabbit
Immunogen Peptide sequence around aa. 392~396 (E-D-S-G-D) derived from Human HDAC2.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3066
Target Species Human
Target Species Murine
Target Species Rat