Please login to your online account to display your discounted pricing

anti-Histidase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227348

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227348

  • / Each
 

Description & Specifications

Specifications

Antigen Histidase
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Host Species Rabbit
Immunogen This antibody was raised against His
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3034
Target Species Human