missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Histidase, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was raised against His
Antigen Histidase
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227348 Novus Biologicals Canada
NBP227348
0.1mg Each for $255.80
NBP227348SS Novus Biologicals Canada
NBP227348SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Histidase
Purified
RUO
3034
0.5mg/mL
This antibody was raised against His
Polyclonal
Primary
Western Blot
100μg antibody in 200μL PBS containing 0.05% BSA.