Please login to your online account to display your discounted pricing

anti-HIPK3, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 850-900 of human HIPK3 was used as the immunogen.
Antigen HIPK3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224521SS

 
novus biologicals canada
NBP224521SS
Unlabeled 0.025mg Each for $139.00

NBP224521

 
novus biologicals canada
NBP224521
Unlabeled 0.1mg Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 850-900 of human HIPK3 was used as the immunogen.
Antigen HIPK3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_c) and chimpanzee, 94% homologous in orangutan and rhesus monkey, 83% homologous in horse and mouse, 77% homologous in cow and 47% homologous in rat.
Gene Accession No. Q9H422
Gene ID (Entrez) 10114
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL