Please login to your online account to display your discounted pricing

anti-HINFP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056628

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056628

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 25988
Primary or Secondary Primary
Immunogen 180-194
Applications Western Blotting
Formulation PBS containing 0.2% gelatin
Test Specificity In MOLT-4, a 64 kDa band is observed.
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q9BQA5
Conjugate Unlabeled
Antigen HINFP
Host Species Rabbit
Format Antiserum
Quantity 0.2mL
Regulatory Status RUO
Cross Reactivity Human