Please login to your online account to display your discounted pricing

anti-HINFP, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen 180-194
Antigen HINFP
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056628T

 
novus biologicals canada
NB10056628SS
Unlabeled 0.05mL Each for $139.00

NB10056628

 
novus biologicals canada
NB10056628
Unlabeled 0.2mL Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 180-194
Antigen HINFP
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In MOLT-4, a 64 kDa band is observed.
Gene Accession No. Q9BQA5
Gene ID (Entrez) 25988
Formulation PBS containing 0.2% gelatin