Please login to your online account to display your discounted pricing

anti-HIF-3 alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002287SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002287SS

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mL
Conjugate Unlabeled
Gene Accession No. Q9Y2N7
Immunogen Bacterially expressed human HIF-3 alpha (N-terminus). [UniProt No. Q9Y2N7]
Gene ID (Entrez) 64344
Concentration 1mg/mL
Regulatory Status RUO
Antigen HIF-3 alpha
Format Affinity Purified
Applications Western Blotting
Host Species Rabbit
Primary or Secondary Primary
Cross Reactivity Mouse