Please login to your online account to display your discounted pricing

anti-HIF-3 alpha, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human HIF-3 alpha (within residues 250-350). [Swiss-Prot No. Q66K72]
Antigen HIF-3 alpha
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002529

 
novus biologicals canada
NB1002529
Unlabeled 0.1mL Each for $329.00

NB1002529SS

 
novus biologicals canada
NB1002529SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of human HIF-3 alpha (within residues 250-350). [Swiss-Prot No. Q66K72]
Antigen HIF-3 alpha
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q9Y2N7
Gene ID (Entrez) 64344
Formulation PBS