Please login to your online account to display your discounted pricing

anti-HIF-3 alpha, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002529

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002529

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9Y2N7
Conjugate Unlabeled
Format Purified
Primary or Secondary Primary
Formulation PBS
Gene ID (Entrez) 64344
Immunogen A synthetic peptide made to an internal region of human HIF-3 alpha (within residues 250-350). [Swiss-Prot No. Q66K72]
Cross Reactivity Human
Cross Reactivity Mouse
Antigen HIF-3 alpha
Quantity 0.1mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Regulatory Status RUO