Please login to your online account to display your discounted pricing

anti-HIF-1 alpha, Polyclonal, Novus Biologicals™

$144.63 - $332.66

Specifications

Host Species Rabbit
Immunogen Genomic sequence made to an internal portion of human HIF-1 alpha (within residues 400-550). [Swiss-Prot No. Q16665]
Antigen HIF-1 alpha
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP102160SS

 
novus biologicals canada
NBP102160SS
Unlabeled 0.025mL Each for $144.63

NBP102160

 
novus biologicals canada
NBP102160
Unlabeled 0.1mL Each for $332.66
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Genomic sequence made to an internal portion of human HIF-1 alpha (within residues 400-550). [Swiss-Prot No. Q16665]
Antigen HIF-1 alpha
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blotting
Test Specificity Human. Reactivity with mouse and porcine reported by a customer review. Predicted to react with monkey based on 100% sequence homology. Immunogen sequence has 96% homology to bovine, 95% to porcine, 94% to rabbit, 92% to rat, 93% to mouse isoform 2, 83% to mouse isoform 1, and 89% to chicken.
Gene Accession No. Q16665
Gene ID (Entrez) 3091
Formulation PBS, 0.1% BSA, 50% glycerol
Concentration 0.2mg/mL