Please login to your online account to display your discounted pricing

anti-HIF-1 alpha (Exon 9), Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human HIF-1 alpha protein (within residues 350-400). [Swiss-Prot No. Q16665]
Antigen HIF-1 alpha (Exon 9)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP147178

 
novus biologicals canada
NBP147178
Unlabeled 0.1mL Each for $309.05

NBP147178SS

 
novus biologicals canada
NBP147178SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human HIF-1 alpha protein (within residues 350-400). [Swiss-Prot No. Q16665]
Antigen HIF-1 alpha (Exon 9)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Exon 9 of HIF-1 alpha. Human. Western blot testing was performed on the full length human recombinant protein. Predicted to react with mouse, rat, bovine and porcine based on 100% sequence homology.
Gene Accession No. Q16665
Gene ID (Entrez) 3091
Formulation PBS, 30% glycerol
Concentration 1mg/mL