Please login to your online account to display your discounted pricing

anti-HIF-1 alpha (Exon 12), Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human HIF-1 alpha protein (within residues 550-600). [Swiss-Prot No. Q16665]
Antigen HIF-1 alpha (Exon 12)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP147180SS

 
novus biologicals canada
NBP147180SS
Unlabeled 0.025mL Each for $109.00

NBP147180

 
novus biologicals canada
NBP147180
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human HIF-1 alpha protein (within residues 550-600). [Swiss-Prot No. Q16665]
Antigen HIF-1 alpha (Exon 12)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Exon 12 of HIF-1 alpha. Human. Western blot testing was performed on the full length human recombinant protein. Predicted to react with mouse, rat, bovine and porcine based on 100% sequence homology. Immunogen sequence has 84% homology to Xenopus.
Gene Accession No. Q16665
Gene ID (Entrez) 3091
Formulation PBS, 30% glycerol
Concentration 1mg/mL