Please login to your online account to display your discounted pricing

anti-HIF-1 alpha (Exon 10), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP147179

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP147179

  • / Each
 

Description & Specifications

Specifications

Antigen HIF-1 alpha (Exon 10)
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q16665
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human HIF-1 alpha protein (within residues 400-450). [Swiss-Prot No. Q16665]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 3091
Test Specificity Exon 10 of HIF-1 alpha. Human. Western blot testing was performed on the full length human recombinant protein. Immunogen sequence has 95% homology to bovine, 90% to porcine and 85% homology to mouse and rat.
Target Species Human