Please login to your online account to display your discounted pricing

anti-HES7, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen RAENRDGPkMLkPLVEkRc (aa 6-23)
Antigen HES7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227417SS

 
novus biologicals canada
NBP227417SS
Unlabeled 0.025mg Each for $109.00

NBP227417

 
novus biologicals canada
NBP227417
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen RAENRDGPkMLkPLVEkRc (aa 6-23)
Antigen HES7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene ID (Entrez) 84667