Please login to your online account to display your discounted pricing

anti-HES2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227110

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227110

  • / Each
 

Description & Specifications

Specifications

Antigen HES2
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen 8 GDAAELRkSLkPLLEkRR 25
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 54626
Test Specificity Chimpanzee, Human, Mouse, Rat.
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat