Please login to your online account to display your discounted pricing

anti-HE4/WFDC2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227294SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227294SS

  • / Each
 

Description & Specifications

Specifications

Antigen HE4/WFDC2
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg HE4 antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. Q14508
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of (human) HE4 was used as the immunogen for the antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 10406
Target Species Human