Please login to your online account to display your discounted pricing

anti-HE4/WFDC2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227294

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227294

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 10406
Gene Accession No. Q14508
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Primary or Secondary Primary
Formulation 0.1mg HE4 antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL
Regulatory Status RUO
Antigen HE4/WFDC2
Immunogen A portion of amino acids 1-50 of (human) HE4 was used as the immunogen for the antibody.
Format Purified
Cross Reactivity Human
Quantity 0.1mg