missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-HE4/WFDC2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of (human) HE4 was used as the immunogen for the antibody.
Antigen HE4/WFDC2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227294SS Novus Biologicals Canada
NBP227294SS
0.025mg Each for $109.00
NBP227294 Novus Biologicals Canada
NBP227294
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
HE4/WFDC2
Purified
RUO
Q14508
0.1mg HE4 antibody in 0.2mL PBS containing 0.05% BSA.
A portion of amino acids 1-50 of (human) HE4 was used as the immunogen for the antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin)
10406
0.5mg/mL