Please login to your online account to display your discounted pricing

anti-HDLBP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224769SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224769SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3069
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q00341
Concentration 0.5mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, cow, zebrafish, pig, Xenopus, rhesus monkey and horse. Fibroblast growth factor receptor proteins share a high homology in the regions surrounding the phosphorylation sites. The immunogen sequence used is 100% homologous in FGFR1-4 and each receptor may have multiple isoforms.
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Antigen HDLBP
Regulatory Status RUO
Format Purified
Immunogen A portion of amino acids 500-550 and 900-950 of human Vigilin was used as the immunogen.
Quantity 0.025mg
Isotype IgG
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Applications Western Blotting