Please login to your online account to display your discounted pricing

anti-HDLBP, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 500-550 and 900-950 of human Vigilin was used as the immunogen.
Antigen HDLBP
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224769

 
novus biologicals canada
NBP224769
Unlabeled 0.1mg Each for $257.41

NBP224769SS

 
novus biologicals canada
NBP224769SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 500-550 and 900-950 of human Vigilin was used as the immunogen.
Antigen HDLBP
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, cow, zebrafish, pig, Xenopus, rhesus monkey and horse. Fibroblast growth factor receptor proteins share a high homology in the regions surrounding the phosphorylation sites. The immunogen sequence used is 100% homologous in FGFR1-4 and each receptor may have multiple isoforms.
Gene Accession No. Q00341
Gene ID (Entrez) 3069
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL