Please login to your online account to display your discounted pricing

anti-HDAC9, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human HDAC-9 was used as the immunogen.
Antigen HDAC9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203993SS

 
novus biologicals canada
NBP203993SS
Unlabeled 0.025mg Each for $139.00

NBP203993

 
novus biologicals canada
NBP203993
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human HDAC-9 was used as the immunogen.
Antigen HDAC9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse, rat, mouse, opossum, chimpanzee, rhesus monkey, orangutan, dog and cow and 94% homologous in chicken.
Gene Accession No. AAI11736.1
Gene ID (Entrez) 9734
Formulation 100μg in 200μL PBS containing 0.05% BSA